Merck
Low-grade neuroinflammation due to chronic sleep deprivation results in anxiety and learning and memory impairments.
Molecular and cellular biochemistry (2018-03-20)
Shaffi Manchanda, Harpal Singh, Taranjeet Kaur, Gurcharan Kaur
RÉSUMÉ
Chronic sleep loss/fragmentation prevalent in the current 24/7 society is associated with irreversible consequences on health and overall wellbeing. Various studies have well documented the ill effects of acute sleep loss on cognitive functions of individuals; however, the underlying mechanism behind the chronic sleep loss is yet to be explored. The present study was aimed to investigate whether chronic sleep deprivation (CSD) triggers anxiety-like behaviour and memory decline in male Wistar rats. Rats were sleep deprived by placing them over slowly rotating drum (2 rpm) for 18 h (between 4 pm and 10 am) followed by 6 h of recovery sleep for 21 consecutive days. Post CSD regimen, rats were subjected to behavioural tests such as elevated plus maze (EPM), Novel Object Recognition (NOR) and Rotarod performance test and then sacrificed to remove brain for further molecular studies. The study demonstrated that CSD rats showed anxiogenic behaviour along with recognition memory decline compared to control rats. CSD rats further showed elevated levels of inflammatory cytokines (TNFα, IL-1β) along with activation of NFκB and AP1 transcription factors in hippocampus and piriform cortex (PC) regions of brain. These observations were also accompanied by enhanced expression of GFAP and Iba1 in the two brain regions. The data suggest that CSD triggered low-grade neuroinflammation which caused anxiogenic response and recognition memory impairment. The study provides preliminary leads to further explore the role of astrocytes/microglial cells and inflammatory cytokines in mediating these neurobehavioural consequences of chronic sleep loss and to develop effective interventions to combat them.