Merck
  • Alda-1, an aldehyde dehydrogenase-2 agonist, causes deterioration in renal functions following ischemia-reperfusion injury due to crystalline nephropathy.
Alda-1, an aldehyde dehydrogenase-2 agonist, causes deterioration in renal functions following ischemia-reperfusion injury due to crystalline nephropathy.
Drug development research (2018-10-07)
Fayez T Hammad, Suhail Al-Salam, Priya Yuvaraju, Loay Lubbad
RÉSUMÉ
Renal ischemia-reperfusion injury (IRI) induces the production of aldehydes which are detoxified by aldehyde dehydrogenases (ALDHs). Alda-1 is a selective ALDH2 agonist and its protective effect was demonstrated in several conditions. The effect of Alda-1 on the kidney or on renal IRI was not investigated. We investigated the effect of Alda-1 on the renal dysfunction following IRI. Wistar rats underwent left IRI for 40 min. Group-Alda (n = 11) received Alda-1 starting 24 h before IRI and continued for 7 days thereafter when renal functions were measured. Group-Vx (n = 11) underwent similar protocol but received the dissolvent. Alda-1 did not affect renal blood flow or glomerular filtration rate in the left ischemic kidney in Group-Alda compared to Group-Vx (3.05 ± 0.50 vs. 3.53 ± 0.70, and 0.40 ± 0.06 vs. 0.51 ± 0.08, respectively, p > .05 for both). However, left renal fractional sodium excretion was higher in Group-Alda (2.80 ± 0.43 vs. 1.37 ± 0.36, p = .02). Alda-1 also adversely affected the gene expressions of kidney injury molecule-1 and neutrophil gelatinase-associated lipocalin (217 ± 38 vs. 99 ± 13 and 49 ± 13 vs. 20 ± 5, respectively, p < .05 for both) and the alterations in tumor necrosis factor-α, transforming growth factor-β1, plasminogen activator inhibitor-1, fibronectin 1 and p53 (4.4 ± 0.9 vs. 2.1 ± 0.3, 1.5 ± 0.1 vs. 1.1 ± 0.1, 30.0 ± 2.7 vs. 11.7 ± 2.3, 3.6 ± 0.4 vs. 2.1 ± 0.2 and 1.3 ± 0.1 vs. 0.9 ± 0.07, respectively, p ≤ .05 for all). This was associated with intratubular crystal deposition suggestive of crystalline nephropathy. Alda-1 exacerbated the IRI-induced renal tubular dysfunction and alterations in markers of acute kidney injury, biomarkers of inflammation, fibrosis and apoptosis and this was associated with intratubular crystal deposition suggestive of crystalline nephropathy.