Merck
Overexpression of microRNA-98 inhibits cell proliferation and promotes cell apoptosis via claudin-1 in human colorectal carcinoma.
Journal of cellular biochemistry (2018-12-07)
Yi-Feng Zheng, Jie Luo, Guo-Lian Gan, Wei Li
RÉSUMÉ
Colorectal carcinoma (CRC) is a major cause of cancer-related deaths worldwide, and investigations on novel targets are imperative. MiR-98 has been reported to act as a tumor suppressor in several cancers. To evaluate miR-98 as a novel anticancer molecule for CRC, examinations to validate whether miR-98 conferred an inhibiting effect on proliferation, migration, and invasion were performed. The microarray-based gene expression profiling involving CRC was used to identify the differentially expressed genes. The potential relationship between miR-98 and CLDN1 was analyzed by cell experimentation. Then, the CRC cells were transfected with miR-98 mimic or miR-98 inhibitor to investigate the potential effect of miR-98 overexpression and depletion on CRC cell proliferation, migration, invasion, and apoptosis. The expressions of CLDN1, Bcl-2 associated protein x (Bax), runt-related transcription factor 3 (RUNX3), B-cell lymphoma 2 (Bcl-2), C-myc, and proliferating cell nuclear antigen (PCNA) were determined. The downregulated miR-98 along with an upregulated CLDN1 was observed in CRC, in which miR-98 could target to regulate CLDN1. The overexpression of miR-98 or silencing of CLDN1 was shown to increase the expression of Bax and RUNX3 along with promoted cell apoptosis and arrested cells in G1 phase, while decreasing the expression of CLDN1, Bcl-2, C-myc, and PCNA with suppressed proliferation, migration, and invasion. Collectively, the current study supports the notion that miR-98 plays an inhibitory role in human CRC cell proliferation, migration, and invasion and act as a contributor for cell apoptosis by downregulating CLDN1. The current study highlights a potential future strategy to help prevent the development of CRC.