Merck
  • Paired Utility of Aza-Amino Acyl Proline and Indolizidinone Amino Acid Residues for Peptide Mimicry: Conception of Prostaglandin F2α Receptor Allosteric Modulators That Delay Preterm Birth.
Paired Utility of Aza-Amino Acyl Proline and Indolizidinone Amino Acid Residues for Peptide Mimicry: Conception of Prostaglandin F2α Receptor Allosteric Modulators That Delay Preterm Birth.
Journal of medicinal chemistry (2019-04-02)
Fatemeh M Mir, N D Prasad Atmuri, Carine B Bourguet, Jennifer Rodon Fores, Xin Hou, Sylvain Chemtob, William D Lubell
RÉSUMÉ
Peptide mimicry employing a combination of aza-amino acyl proline and indolizidinone residues has been used to develop allosteric modulators of the prostaglandin F2α receptor. The systematic study of the N-terminal phenylacetyl moiety and the conformation and side chain functions of the central turn dipeptide residue has demonstrated the sensitive relationships between modulator activity and topology. Examination of aza-Gly-Pro and aza-Phe-Pro analogs 2a and 2b in a murine preterm labor model featuring treatment with lipopolysaccharide demonstrated their capacity to extend significantly (>20 h) the average time of delivery offering new prototypes for delaying premature birth.