Merck
Old Molecule, New Chemistry: Exploring Silicon Phthalocyanines as Emerging N-Type Materials in Organic Electronics.
Materials (Basel, Switzerland) (2019-04-27)
Nathan J Yutronkie, Trevor M Grant, Owen A Melville, Benoît H Lessard, Jaclyn L Brusso
RÉSUMÉ
Efficient synthesis of silicon phthalocyanines (SiPc) eliminating the strenuous reaction conditions and hazardous reagents required by classical methods is described. Implementation into organic thin-film transistors (OTFTs) affords average electron field-effect mobility of 3.1 × 10-3 cm2 V-1 s-1 and threshold voltage of 25.6 V for all synthetic routes. These results demonstrate that our novel chemistry can lead to high performing SiPc-based n-type OTFTs.