Merck
High calpain-1 expression predicts a poor clinical outcome and contributes to tumor progression in pancreatic cancer patients.
Clinical & translational oncology : official publication of the Federation of Spanish Oncology Societies and of the National Cancer Institute of Mexico (2018-12-20)
L M Yu, Y S Zhu, C Z Xu, L L Zhou, Z X Xue, Z Z Cai
RÉSUMÉ
Pancreatic cancer (PC) is a highly aggressive and metastatic disease, with an elevated mortality rate. It is, therefore, crucial to assess factors affecting the prognosis of PC patients. Meanwhile, calpain-1 is associated with malignant tumor progression and metastasis. Thus, it is meaningful to evaluate the relationship between calpain-1 and PC. Calpain-1 protein expression was assessed by immunohistochemistry in 96 pancreatic cancer samples and paired adjacent non-cancerous specimens. In addition, calpain-1 protein levels were assessed in six PC cell lines by western blot (WB). Next, PC cells were transfected with calpain-1 siRNA, and silencing was confirmed by WB. Finally, cell proliferation, colony formation, migration and invasion assays, and cell apoptosis analysis were performed to examine the effects of calpain-1 knockdown on proliferation, growth, apoptosis, migration, and invasion in PC cells. The results showed that calpain-1 was overexpressed in PC tissues and cells. Meanwhile, calpain-1 overexpression was associated with tumor site (P = 0.029), metastasis (P = 0.000), and TNM stage (P = 0.000), but showed no associations with histological grade (P = 0.396), age (P = 0.809), sex (P = 1.000), and lesion size (P = 0.679). The Kaplan-Meier method demonstrated that the low calpain-1 expression group had increased overall survival (OS) compared with patients expressing high calpain-1 levels (28.7 ± 4.1 vs. 17.0 ± 2.3 months) (P = 0.005). Besides, calpain-1 in PC cells was successfully silenced by liposome-mediated RNA interference, resulting in reduced cell growth, invasion, and metastasis in PC cells, with no effect on apoptosis. The above findings suggest that calpain-1 should be considered a potential biomarker for PC prognosis and therapy.