Merck
  • Amphiregulin potentiates airway inflammation and mucus hypersecretion induced by urban particulate matter via the EGFR-PI3Kα-AKT/ERK pathway.
Amphiregulin potentiates airway inflammation and mucus hypersecretion induced by urban particulate matter via the EGFR-PI3Kα-AKT/ERK pathway.
Cellular signalling (2018-10-07)
Jian Wang, Mengchan Zhu, Linlin Wang, Cuicui Chen, Yuanlin Song
RÉSUMÉ
Ambient particulate matter (PM) promotes the development and exacerbation of chronic respiratory diseases, including chronic obstructive pulmonary disease (COPD) and asthma, by increasing inflammation and mucus hypersecretion. However, the biological mechanisms underlying PM-induced airway inflammation and mucus hypersecretion remain unclear. Amphiregulin (AREG) is an important ligand for epidermal growth factor receptor (EGFR) and participates in the regulation of several biological functions. Here, the PM-exposed human bronchial epithelial cell (HBEC) model was used to define the role of AREG in PM-induced inflammation and mucus hypersecretion and its related signaling pathways. The expression of AREG was significantly increased in a dose-dependent manner in HBECs subjected to PM exposure. Moreover, PM could induce inflammation and mucus hypersecretion by upregulating the expression of IL-1α, IL-1β, and Muc-5ac in HBECs. The EGFR, AKT, and ERK signaling pathways were also activated in a time- and dose-dependent manner. The AREG siRNA markedly attenuated PM-induced inflammation and mucus hypersecretion, and activation of the EGFR-AKT/ERK pathway. Exogenous AREG significantly increased the expression of IL-1α, IL-1β, and Muc-5ac, and induced activation of the EGFR-AKT/ERK pathway in HBECs. Further, under PM exposure, exogenous AREG significantly potentiated PM-induced inflammation and mucus hypersecretion, and activation of the EGFR-AKT/ERK pathway. Tumor-necrosis factor-alpha converting enzyme (TACE) and EGFR specific inhibitor pretreatment showed that AREG was secreted by TACE-mediated cleavage to regulate PM-induced inflammation and mucus hypersecretion by binding to the EGFR. Moreover, according to the inhibitory effect of specific inhibitors of the class I PI3K isoforms, AKT and ERK, PM-induced inflammation and mucus hypersecretion was regulated by PI3Kα activation and its downstream AKT and ERK pathways. This study strongly suggests the adverse effect of AREG in PM-induced inflammation and mucus hypersecretion via the EGFR-PI3Kα-AKT/ERK pathway. These findings contribute to a better understanding of the biological mechanisms underlying exacerbation of chronic respiratory diseases induced by PM exposure.