Merck
Optimizing ligand structure for low-loading and fast catalysis for alkynyl-alcohol and -amine cyclization.
Dalton transactions (Cambridge, England : 2003) (2019-05-16)
James M Stubbs, Benjamin J Bridge, Johanna M Blacquiere
RÉSUMÉ
A series of [Ru(Cp/Cp*)(PR2NR'2)(MeCN)]PF6 complexes were prepared, in which the steric and electronic properties of the primary coordination sphere were varied (R = Ph, t-Bu, Bn; and Cp vs. Cp*). These complexes were catalytically active in the cyclization of alkyne substrates with an intramolecular nucleophile (amine or alcohol) to produce 5- and 6-membered heterocycles. The effect of the 1° coordination sphere structure on catalyst performance was evaluated. Steric bulk around the metal centre was a key feature to achieve rapid catalysis at low temperatures. The catalyst [Ru(Cp)(Pt-Bu2NPh2)(MeCN)]PF6 gave a turnover number that was >1 order of magnitude more active than previous catalysts in the cyclization of the benchmark substrate 2-ethynylaniline. This catalyst is tolerant of a diversity of functional groups and is competent at the formation of various substituted indoles.