Merck
Effects of cyclophosphamide on the phenotypes and functions of THP-1 cells.
Environmental toxicology and pharmacology (2019-06-16)
Youliang Zhao, Changfu Hao, Ruonan Zhai, Lei Bao, Di Wang, Yiping Li, Xinghao Yu, Ruoxuan Huang, Wu Yao
RÉSUMÉ
Early and accurate evaluation of immunotoxicity is crucial. However, there are few in vitro models for immunosuppressive evaluation. THP-1 cells has long been used for in vitro sensitivity evaluation. Whether it can be used for immunosuppressive evaluation remains unclear. In this study, effects of immunosuppressant cyclophosphamide (CY) on THP-1 cells were observed while 2, 4-Dinitrochlorobenzene (DNCB) was used as a control. The phenotypes of THP-1 cells, the ability to activate naïve T cells, intracellular reactive oxygen species (ROS) level, gene markers, phagocytic ability and cell apoptosis were detected after THP-1 cells being exposed to different concentrations of CY and DNCB. Both CY and DNCB were able to activate THP-1 cells, but there were a lot of differences in their effects on THP-1 cells, such as the changes in phenotypes, in the ability to activate naïve T cells, in ROS production and in marker gene expression. Firstly, CY down-regulated the expression of CD86 on THP-1 cells while DNCB up-regulated its expression. Secondly, the ability of THP-1 cells to activate naïve T cells was enhanced by CY and suppressed by DNCB. Thirdly, CY raised rapid and transient elevation of ROS level in THP-1 cells, while the effects of DNCB were slower and longer-lasting. Finally, only CY could lead to an increase in heme oxygenase 1 (HMOX1) expression. Taken all these results into account, we suggested that THP-1 cell line possesses the potency to be an in vitro model of immunosuppressive evaluation. And the surface molecule CD86, the ability to activate naïve T cells, the ROS production and the gene marker HMOX1 of THP-1 cells are promising markers.