Merck
Stereoselective isomerisation of N-allyl aziridines into geometrically stable Z enamines by using rhodium hydride catalysis.
Chemistry (Weinheim an der Bergstrasse, Germany) (2007-11-10)
Derek S Tsang, Sharon Yang, France-Aimée Alphonse, Andrei K Yudin
RÉSUMÉ
In the presence of rhodium(I) hydride catalysts, tertiary N-allylamines are known to isomerise into E enamines. In contrast, we have recently found that N-allylaziridines isomerise to form Z enamines. On the basis of literature data, the most likely mechanism of isomerisation would involve a rhodium hydride addition/beta-hydride elimination sequence. We show that the observed selectivity cannot be adequately explained by this pathway and is more consistent with initial CH-activation followed by rearrangement to form a five-membered cyclometallated rhodium intermediate. This intermediate subsequently undergoes reductive elimination to form a C--H bond. The resulting geometrically stable Z enamines are useful building blocks for stereoselective synthesis.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Vinylboronic anhydride pyridine complex, 95%