Merck
Polyproline Tri-Helix Macrocycles as Nanosized Scaffolds to Control Ligand Patterns for Selective Protein Oligomer Interactions.
Small (Weinheim an der Bergstrasse, Germany) (2019-04-13)
Cin-Hao Lin, Hsin-Chuan Wen, Cheng-Chin Chiang, Jen-Sheng Huang, Yunching Chen, Sheng-Kai Wang
RÉSUMÉ
Multivalent ligand-receptor interactions play essential roles in biological recognition and signaling. As the receptor arrangement on the cell surface can alter the outcome of cell signaling and also provide spatial specificity for ligand binding, controlling the presentation of ligands has become a promising strategy to manipulate or selectively target protein receptors. The lack of adjustable universal tools to control ligand positions at the size of a few nanometers has prompted the development of polyproline tri-helix macrocycles as scaffolds to present ligands in designated patterns. Model lectin Helix pomatia agglutinin has shown selectivity toward the matching GalNAc ligand pattern matching its binding sites arrangement. The GalNAc pattern selectivity is also observed on intact asialoglycoprotein receptor oligomer on human hepatoma cells showing the pattern-selective interaction can be achieved not only on isolated protein oligomers but also the receptors arranged on the cell surface. As the scaffold design allows convenient creation of versatile ligand patterns, it can be expected as a promising tool to probe the arrangement of receptors on the cell surface and as nanomedicine to manipulate signaling or cell recognition.