Merck
Could the porous chitosan-based composite materials have a chance to a "NEW LIFE" after Cu(II) ion binding?
International journal of biological macromolecules (2019-03-13)
Maria Marinela Lazar, Ionel Adrian Dinu, Mihaela Silion, Ecaterina Stela Dragan, Maria Valentina Dinu
RÉSUMÉ
Currently, biosorption is considered a leading-edge environmentally-friendly method for the low-cost remediation of wastewaters contaminated with metal ions. However, the safe disposal of metal-loaded biosorbents is still a challenging issue. In this context, our major objective was to explore the possibility of "waste minimization" by reusing the metal-loaded biosorbents in further environmental applications, particularly into the oxidative catalysis of dyes. Thus, the decolourisation efficiency (DE) of Methyl Orange (MO) in aqueous solutions under ambient light using copper-imprinted chitosan-based composites in comparison to non-imprinted ones was investigated in this work. The MO degradation was established first in the absence of any co-catalyst, when a DE value of 95.3% was achieved by the ion-imprinted catalysts within 360 min of reaction, compared to only 67.4% attained by the non-imprinted ones. Under Fenton-like conditions, the apparent degradation rate constant was seventy times higher, the DE increasing within 40 min to about 98.6%, and 70.5% respectively, whereas the content of co-catalyst (H2O2) was significantly lowered compared to other reported studies. The straightforward preparation of copper-loaded composites, along with their excellent stability and high efficiency even after four consecutive reaction runs support our ion-imprinted systems as potential catalysts for dye removal by oxidative decolourisation treatments.