Merck
  • Hyperlipoproteinemia in Nagase analbuminemic rats: effects of pravastatin on plasma (apo)lipoproteins and lecithin:cholesterol acyltransferase activity.
Hyperlipoproteinemia in Nagase analbuminemic rats: effects of pravastatin on plasma (apo)lipoproteins and lecithin:cholesterol acyltransferase activity.
Journal of lipid research (1991-11-01)
A Van Tol, E H Jansen, H A Koomans, J A Joles
RÉSUMÉ
The present study demonstrates very high levels of plasma lipids and high density lipoprotein (HDL) apolipoproteins (apoA-I and apoE) in female Nagase analbuminemic rats (NAR) fed a semi-synthetic diet in order to further increase the hyperlipidemia present in this strain. Plasma apoB-containing lipoproteins (very low, intermediate, and low density lipoproteins) were also elevated in NAR. Plasma cholesterol was mainly present in lipoprotein particles with a density between 1.02 and 1.12 g/ml. Separation of lipoprotein classes by gel filtration showed that the major cholesterol-carrying lipoprotein fractions in NAR plasma are apoE-rich HDL and apoA-I-rich HDL. The high HDL levels in NAR are explained, at least partly, by the two- to threefold elevated activity of plasma lecithin:cholesterol acyltransferase (LCAT). The lysophosphatidylcholine generated in the LCAT reaction, as well as plasma free fatty acids, are bound to lipoproteins in NAR plasma. A study was carried out to determine whether the elevated LDL and aopoE-rich HDL levels could be corrected by administration of the HMG-CoA reductase inhibitor pravastatin (at a dose of 1 mg/kg per day). Pravastatin treatment results in a 43% decrease in plasma triglycerides in NAR, but not in Sprague-Dawley (SDR) rats, and had no significant effect on plasma total cholesterol, phospholipids apolipoproteins A-I, A-IV, B, or E, as well as on plasma LCAT activity levels in NAR or SDR.