- NAD-dependent inhibition of the NAD-glycohydrolase activity in A549 cells.
NAD-dependent inhibition of the NAD-glycohydrolase activity in A549 cells.
NAD glycohydrolases are enzymes that catalyze the hydrolysis of NAD to produce ADP-ribose and nicotinamide. Regulation of these enzymes has not been fully elucidated. We have identified a NAD-glycohydrolase activity associated with the outer surface of the plasma membrane in human lung epithelial cell line A549. This activity is negatively regulated by its substrate beta-NAD but not by alpha-NAD. Partial restoration of NADase activity after incubation of the cells with arginine or histidine, known ADP-ribose acceptors, suggests that inhibition be regulated by ADP-ribosylation. A549 do not undergo to apoptosis upon NAD treatment indicating that this effect be likely mediated by a cellular component(s) lacking in epithelial cells.