- What has senescence got to do with cancer?
RÉSUMÉ
Cancer therapeutics are primarily thought to work by inducing apoptosis in tumor cells. However, various tumor suppressors and oncogenes have been shown to regulate senescence in normal cells, and senescence bypass appears to be an important step in the development of cancer. Cellular senescence limits the replicative capacity of cells, thus preventing the proliferation of cells that are at different stages of malignancy. A recent body of evidence suggests that induction of senescence can be exploited as a basis for cancer therapy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Dosage de la sénescence cellulaire, Cellular Senescence Assay Kits provide all the reagents required to efficiently detect SA-β-gal activity at pH 6.0 in cultured cells & tissue sections.