Merck
  • Highly Fluorinated Ir(III)-2,2':6',2″-Terpyridine-Phenylpyridine-X Complexes via Selective C-F Activation: Robust Photocatalysts for Solar Fuel Generation and Photoredox Catalysis.
Highly Fluorinated Ir(III)-2,2':6',2″-Terpyridine-Phenylpyridine-X Complexes via Selective C-F Activation: Robust Photocatalysts for Solar Fuel Generation and Photoredox Catalysis.
Journal of the American Chemical Society (2016-07-09)
Jonathan A Porras, Isaac N Mills, Wesley J Transue, Stefan Bernhard
RÉSUMÉ
A series of fluorinated Ir(III)-terpyridine-phenylpyridine-X (X = anionic monodentate ligand) complexes were synthesized by selective C-F activation, whereby perfluorinated phenylpyridines were readily complexed. The combination of fluorinated phenylpyridine ligands with an electron-rich tri-tert-butyl terpyridine ligand generates a "push-pull" force on the electrons upon excitation, imparting significant enhancements to the stability, electrochemical, and photophysical properties of the complexes. Application of the complexes as photosensitizers for photocatalytic generation of hydrogen from water and as redox photocatalysts for decarboxylative fluorination of several carboxylic acids showcases the performance of the complexes in highly coordinating solvents, in some cases exceeding that of the leading photosensitizers. Changes in the photophysical properties and the nature of the excited states are observed as the compounds increase in fluorination as well as upon exchange of the ancillary chloride ligand to a cyanide. These changes in the excited states have been corroborated using density functional theory modeling.