Merck
The first tandem, all-exciplex-based WOLED.
Scientific reports (2014-06-05)
Wen-Yi Hung, Guan-Cheng Fang, Shih-Wei Lin, Shuo-Hsien Cheng, Ken-Tsung Wong, Ting-Yi Kuo, Pi-Tai Chou
RÉSUMÉ
Exploiting our recently developed bilayer interface methodology, together with a new wide energy-gap, low LUMO acceptor (A) and the designated donor (D) layers, we succeeded in fabricating an exciplex-based organic light-emitting diode (OLED) systematically tuned from blue to red. Further optimization rendered a record-high blue exciplex OLED with η(ext) of 8%. We then constructed a device structure configured by two parallel blend layers of mCP/PO-T2T and DTAF/PO-T2T, generating blue and yellow exciplex emission, respectively. The resulting device demonstrates for the first time a tandem, all-exciplex-based white-light OLED (WOLED) with excellent efficiencies η(ext): 11.6%, η(c): 27.7 cd A(-1), and η(p): 15.8 ml W(-1) with CIE(0.29, 0.35) and CRI 70.6 that are nearly independent of EL intensity. The tandem architecture and blend-layer D/A (1:1) configuration are two key elements that fully utilize the exciplex delay fluorescence, providing a paragon for the use of low-cost, abundant organic compounds en route to commercial WOLEDs.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
PO-T2T, >=99% (HPLC)