Merck
Rho proteins, mental retardation and the cellular basis of cognition.
Trends in neurosciences (2002-05-10)
Ger J A Ramakers
RÉSUMÉ
For several decades, it has been known that mental retardation (MR) is associated with abnormalities in dendrites and dendritic spines. The recent cloning of seven genes that cause nonspecific MR when mutated provides important insights in the cellular mechanisms that result in the dendritic abnormalities associated with MR. Three of the encoded proteins, oligophrenin 1, PAK3 and alpha PIX, interact directly with Rho GTPases. Rho GTPases are key signaling proteins that integrate extracellular and intracellular signals to orchestrate coordinated changes in the actin cytoskeleton essential for directed neurite outgrowth and the regulation of synaptic connectivity. Although many details of the cell biology of Rho signaling in the CNS are still unclear, a picture is unfolding showing how mutations that alter Rho signaling result in abnormal neuronal connectivity and deficient cognitive functioning in humans. Conversely, these findings illuminate the cellular mechanisms underlying normal cognitive function.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
PAK3 Protein, active, 10 µg, Active, C-terminal 6His-tagged, recombinant, human full length, PAK3. For use in Kinase Assays.