Merck
COMP (cartilage oligomeric matrix protein) is synthesized in ligament, tendon, meniscus, and articular cartilage.
Connective tissue research (2000-11-04)
G Müller, A Michel, E Altenburg
RÉSUMÉ
The presence of cartilage oligomeric matrix protein (COMP) in extracts of ligament, tendon, meniscus, and canine articular cartilage was demonstrated by Western blot analysis using anti-dog COMP antibody. When the tissues were cultured in the presence of [35-S]methionine/cysteine, metabolically labeled COMP was purified from the culture media and from tissue extracts by DEAE-cellulose gel chromatography. SDS-Polyacrylamide gel electrophoresis (SDS-PAGE) followed by autoradiography and immunoblotting under reducing and non-reducing conditions revealed that COMP is synthesized by the cells of these connective tissues. Increased levels of COMP in samples of both synovial fluid and serum of patients with various joint diseases may not only be derived from cartilage but also from ligaments and tendons. COMP is not a highly tissue-specific cartilage molecule.