Merck
  • A versatile catalyst system for Suzuki-Miyaura syntheses of sterically hindered biaryls employing a cyclobutene-1,2-bis(imidazolium) salt.
A versatile catalyst system for Suzuki-Miyaura syntheses of sterically hindered biaryls employing a cyclobutene-1,2-bis(imidazolium) salt.
Chemical communications (Cambridge, England) (2010-04-14)
Andreas Schmidt, Alireza Rahimi
RÉSUMÉ
The catalyst system consisting of 3,3'-(3,4-bis(dichloro-methylene)cyclobut-1-ene-1,2-diyl)bis(1-methyl-1H-imidazolium) bis(tetrafluoroborate), Pd(OAc)(2) and NaOtBu in toluene proved to be very effective for a broad variety of Suzuki-Miyaura reactions at room temperature. It is also suited for the synthesis of sterically hindered compounds including 2,6-di-tert-butyl-2'-substituted biaryls at elevated temperatures.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2-Bromo-1,3-dimethylbenzene, 98%