Merck
TWIK-1/TASK-3 heterodimeric channels contribute to the neurotensin-mediated excitation of hippocampal dentate gyrus granule cells.
Experimental & molecular medicine (2018-11-13)
Jae Hyouk Choi, Oleg Yarishkin, Eunju Kim, Yeonju Bae, Ajung Kim, Seung-Chan Kim, Kanghyun Ryoo, Chang-Hoon Cho, Eun Mi Hwang, Jae-Yong Park
RÉSUMÉ
Two-pore domain K+ (K2P) channels have been shown to modulate neuronal excitability. The physiological role of TWIK-1, the first identified K2P channel, in neuronal cells is largely unknown, and we reported previously that TWIK-1 contributes to the intrinsic excitability of dentate gyrus granule cells (DGGCs) in mice. In the present study, we investigated the coexpression of TWIK-1 and TASK-3, another K2P member, in DGGCs. Immunohistochemical staining data showed that TASK-3 proteins were highly localized in the proximal dendrites and soma of DGGCs, and this localization is similar to the expression pattern of TWIK-1. TWIK-1 was shown to associate with TASK-3 in DGGCs of mouse hippocampus and when both genes were overexpressed in COS-7 cells. shRNA-mediated gene silencing demonstrated that TWIK-1/TASK-3 heterodimeric channels displayed outwardly rectifying currents and contributed to the intrinsic excitability of DGGCs. Neurotensin-neurotensin receptor 1 (NT-NTSR1) signaling triggered the depolarization of DGGCs by inhibiting TWIK-1/TASK-3 heterodimeric channels, causing facilitated excitation of DGGCs. Taken together, our study clearly showed that TWIK-1/TASK-3 heterodimeric channels contribute to the intrinsic excitability of DGGCs and that their activities are regulated by NT-NTSR1 signaling.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Référence
Conditionnement
Disponibilité
Prix
Quantité
Sigma-Aldrich
MISSION® esiRNA, targeting human KCNK9
Référence
Conditionnement
Disponibilité
Prix
Quantité