Merck
Stereoselective synthesis of multisubstituted enamides via rhodium-catalyzed carbozincation of ynamides.
Journal of the American Chemical Society (2009-03-04)
Benoit Gourdet, Hon Wai Lam
RÉSUMÉ
A new rhodium-catalyzed carbozincation of ynamides has been developed, using diorganozinc reagents or functionalized alkylzinc halides. The reactions are highly regio- and stereoselective, allowing access to a wide range of multisubstituted enamides, which are increasingly important building blocks for organic synthesis. Utilization of the alkenylzinc intermediates in further carbon-carbon bond-forming reactions to form trisubstituted enamides is also possible.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Chiralyst P310, Umicore, 99%