Merck
Catalytic C-h/olefin coupling.
Accounts of chemical research (2002-10-16)
Fumitoshi Kakiuchi, Shinji Murai
RÉSUMÉ
The cleavage and addition of ortho C-H bonds in various aromatic compounds such as ketones, esters, imines, imidates, nitrile, and aldehydes to olefins and acetlylenes can be achieved catalytically with the aid of ruthenium catalysts. The reaction is generally highly efficient and useful in synthetic methods. The coordination to the metal center by a heteroatom in directing groups such as carbonyl and imino groups is the key. The reductive elimination to form a C-C bond is the rate-determining step.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Chlorobis(cyclooctene)rhodium(I),dimer, 98%
Le tarif et la disponibilité ne sont pas disponibles actuellement.