Merck
Preparation of magnetic resonance contrast agents activated by beta-galactosidase.
Nature protocols (2008-03-08)
Lauren M Urbanczyk-Pearson, Thomas J Meade
RÉSUMÉ
The preparation of beta-EgadMe and alpha-EgadMe, magnetic resonance (MR) contrast agents that can be used for in vivo detection of beta-galactosidase, is described. Diastereomerically pure beta-EgadMe can be synthesized by kinetically resolving the starting material as described in Step 1. The total time for the preparation of the racemic mixture of beta-EgadMe is about 8 d, and the total time for an diastereomerically resolved agent is about 9 d. The final metallated agent is stable at room temperature as a solid or in aqueous buffer (pH 5.5-10) indefinitely. Diastereomerically pure alpha-EgadMe can be prepared by beginning the synthesis with enantiomerically pure bromopropionic acid. The total time for the preparation of racemic alpha-EgadMe or diastereomerically pure alpha-EgadMe is about 8 d. The final metallated agent is stable at room temperature as a solid or in aqueous buffer (pH 5.5-10) indefinitely.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Oxyde de (±)propylène, analytical standard