Merck
Multi-production of high added market value metabolites from diluted methane emissions via methanotrophic extremophiles.
Bioresource technology (2018-07-22)
S Cantera, I Sánchez-Andrea, R Lebrero, P A García-Encina, Alfons J M Stams, R Muñoz
RÉSUMÉ
This study constitutes the first-proof-of-concept of a methane biorefinery based on the multi-production of high profit margin substances (ectoine, hydroxyectoine, polyhydroxyalkanoates (PHAs) and exopolysaccharides (EPS)) using methane as the sole carbon and energy source. Two bubble column bioreactors were operated under different magnesium concentrations (0.2, 0.02 and 0.002 g L-1) to validate and optimize this innovative strategy for valorization of CH4 emissions. High Mg2+ concentrations promoted the accumulation of ectoine (79.7-94.2 mg g biomass-1), together with high hydroxyectoine yields (up to 13 mg g biomass-1) and EPS concentrations (up to 2.6 g L culture broth-1). Unfortunately, PHA synthesis was almost negligible (14.3 mg L-1) and only found at the lowest Mg2+ concentration tested. Halomonas, Marinobacter, Methylophaga and Methylomicrobium, previously described as ectoine producers, were dominant in both bioreactors, Methylomicrobium being the only described methanotroph. This study encourages further research on CH4 biorefineries capable of creating value out of GHG mitigation.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Hydroxyectoine, ≥95% (HPLC)