Merck
Evaluation of the Edman degradation product of vancomycin bonded to core-shell particles as a new HPLC chiral stationary phase.
Chirality (2018-07-04)
Garrett Hellinghausen, Diego A Lopez, Jauh T Lee, Yadi Wang, Choyce A Weatherly, Abiud E Portillo, Alain Berthod, Daniel W Armstrong
RÉSUMÉ
A modified macrocyclic glycopeptide-based chiral stationary phase (CSP), prepared via Edman degradation of vancomycin, was evaluated as a chiral selector for the first time. Its applicability was compared with other macrocyclic glycopeptide-based CSPs: TeicoShell and VancoShell. In addition, another modified macrocyclic glycopeptide-based CSP, NicoShell, was further examined. Initial evaluation was focused on the complementary behavior with these glycopeptides. A screening procedure was used based on previous work for the enantiomeric separation of 50 chiral compounds including amino acids, pesticides, stimulants, and a variety of pharmaceuticals. Fast and efficient chiral separations resulted by using superficially porous (core-shell) particle supports. Overall, the vancomycin Edman degradation product (EDP) resembled TeicoShell with high enantioselectivity for acidic compounds in the polar ionic mode. The simultaneous enantiomeric separation of 5 racemic profens using liquid chromatography-mass spectrometry with EDP was performed in approximately 3 minutes. Other highlights include simultaneous liquid chromatography separations of rac-amphetamine and rac-methamphetamine with VancoShell, rac-pseudoephedrine and rac-ephedrine with NicoShell, and rac-dichlorprop and rac-haloxyfop with TeicoShell.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Trifluoroacétate d'ammonium, 98%