Merck
A transcription-factor-binding surface of coactivator p300 is required for haematopoiesis.
Nature (2002-10-18)
Lawryn H Kasper, Fayçal Boussouar, Paul A Ney, Carl W Jackson, Jerold Rehg, Jan M van Deursen, Paul K Brindle
RÉSUMÉ
The coactivators CBP (Cre-element binding protein (CREB)-binding protein) and its paralogue p300 are thought to supply adaptor molecule and protein acetyltransferase functions to many transcription factors that regulate gene expression. Normal development requires CBP and p300, and mutations in these genes are found in haematopoietic and epithelial tumours. It is unclear, however, which functions of CBP and p300 are essential in vivo. Here we show that the protein-binding KIX domains of CBP and p300 have nonredundant functions in mice. In mice homozygous for point mutations in the KIX domain of p300 designed to disrupt the binding surface for the transcription factors c-Myb and CREB, multilineage defects occur in haematopoiesis, including anaemia, B-cell deficiency, thymic hypoplasia, megakaryocytosis and thrombocytosis. By contrast, age-matched mice homozygous for identical mutations in the KIX domain of CBP are essentially normal. There is a synergistic genetic interaction between mutations in c-Myb and mutations in the KIX domain of p300, which suggests that the binding of c-Myb to this domain of p300 is crucial for the development and function of megakaryocytes. Thus, conserved domains in two highly related coactivators have contrasting roles in haematopoiesis.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
KAT3A (518-1207), GST tagged human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution