Merck
  • Tailoring of Versatile Surface Morphologies on Hot Dip Galvanized Steel in Wet CO2: Aspects on Formation, Barrier Properties, and Utilization as a Substrate for Coatings.
Tailoring of Versatile Surface Morphologies on Hot Dip Galvanized Steel in Wet CO2: Aspects on Formation, Barrier Properties, and Utilization as a Substrate for Coatings.
ACS applied materials & interfaces (2018-06-06)
Ville Saarimaa, Aaretti Kaleva, Juha-Pekka Nikkanen, Jere Manni, Carl Lange, Teemu Paunikallio, Tero Laihinen, Saara Heinonen, Erkki Levänen, Pasi Väisänen, Antti Markkula
RÉSUMÉ
Zinc carbonate and a mixed-phase zinc carbonate were precipitated selectively on hot dip galvanized steel in the presence of CO2 and water. The zinc carbonate was precipitated as a uniform layer with cubic superficial appearance, while the mixed-phase zinc carbonate was precipitated as nanowires. The distinct structures could be formed separately or as a dual structure with nanowires on the outermost surface. The barrier properties were improved by the both patina forms; a significant increase in surface hydrophobicity was obtained. The dual patina structure was successfully coated with an organic coating, and the intact wet CO2-induced patina with both structures was confirmed within the coating. The formed carbonates can be further converted to zinc oxide by calcination, preserving the delicate structures, which opens a wide range of potential applications for the nanostructured ZnO in a variety of future electronic and optoelectronic devices.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Copper(II) hexafluoroacetylacetonate hydrate