Merck
Role of human immunodeficiency virus type 1-specific protease in core protein maturation and viral infectivity.
Journal of virology (1989-06-01)
C Peng, B K Ho, T W Chang, N T Chang
RÉSUMÉ
It is generally believed that the gag gene product of human immunodeficiency virus type 1 (HIV-1) is processed into several core proteins by a virus-specific protease. We used deletion mutation analysis to study the role of HIV-specific protease in the processing of core proteins and its requirement for viral infectivity. Several mutant genomes with deletions in the protease gene were constructed. A mammalian cell line, COS-M6, transfected with the wild-type viral genome was shown to produce virions containing processed core proteins, while COS-M6 cells transfected with two mutated genomes could express only the core protein precursor, Pr56gag. The wild-type transfectant produced infectious virus; both transfectants expressing the mutated genomes also produced virions, and one of them still retained reverse transcriptase activity. However, the mutant viral particles were devoid of infectivity. Virions with a distinct central core and an electron-dense nucleoid budded out from the plasma membrane of COS-M6 cells transfected with the wild-type genome. In contrast, noninfectious virions that budded either into cytoplasmic vacuoles or out from the plasma membrane of COS-M6 cells transfected with mutant genomes contained ring-shaped nucleoids. These results indicate that the HIV-1 protease plays a role not only in the maturation of the core proteins but also in the assembly of the virus and thus is required for viral infectivity.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
HIV Protease, His tagged,recombinant from HIV-1, recombinant, expressed in E. coli, ≥85% (SDS-PAGE)