Merck
A novel serum protein similar to C1q, produced exclusively in adipocytes.
The Journal of biological chemistry (1995-11-10)
P E Scherer, S Williams, M Fogliano, G Baldini, H F Lodish
RÉSUMÉ
We describe a novel 30-kDa secretory protein, Acrp30 (adipocyte complement-related protein of 30 kDa), that is made exclusively in adipocytes and whose mRNA is induced over 100-fold during adipocyte differentiation. Acrp30 is structurally similar to complement factor C1q and to a hibernation-specific protein isolated from the plasma of Siberian chipmunks; it forms large homo-oligomers that undergo a series of post-translational modifications. Like adipsin, secretion of Acrp30 is enhanced by insulin, and Acrp30 is an abundant serum protein. Acrp30 may be a factor that participates in the delicately balanced system of energy homeostasis involving food intake and carbohydrate and lipid catabolism. Our experiments also further corroborate the existence of an insulin-regulated secretory pathway in adipocytes.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
gAcrp30 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture