Merck
Cloning and characterization of a gene rpg1 encoding polygalacturonase of Rhizopus oryzae.
Mycological research (2005-03-11)
Shigenobu Yoshida, Fumihiko Suzuki, Takao Tsukiboshi, Hirosuke Shinohara
RÉSUMÉ
The polygalacturonase (PG)-encoding gene (rpg1) of Rhizopus oryzae, the causal pathogen of rhizopus rot of mulberry, was cloned and sequenced. PGs were partially purified from incubation mixture of 2% pectin medium and their N-terminal amino acid sequences were determined by a gas-phase protein sequencer. RT-PCR was performed using degenerate primers designed from the amino acid sequences, which resulted in part of a PG-encoding gene being obtained. By 3'-RACE and TAIL-PCR analyses, the entire region of the PG-encoding gene was cloned and sequenced. The structural gene comprised 1199 bp coding for 383 amino acids with a putative signal peptide of 26 amino acids, and the open reading frame was interrupted by single intron of 47 bp. Phylogenetic analysis using the deduced amino acid sequence revealed that R. oryzae RPG1 belonged to a clade consisting of exo-PGs of ascomycete fungi.