Documents
Z659436
Bandelin SONOPULS ultrasonic homogenizer accessories
Indirect processing cup booster BR 30
Synonyme(s) :
high-intensity sonication vessel
About This Item
Code UNSPSC :
41101520
Nomenclature NACRES :
NB.74
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bandelin 7510
Disponibilité
available only in EU
Description générale
For high-intensive irradiation of smallest and sensitive sample volumes, e. g. cleaning of radioactively contaminated seeds, cell disruption or treatment of pathogens and toxic material. No cross-contamination or sample loss. Flow-through cooling liquid avoids rise of temperature