Principaux documents
CLS401577
Corning® Modified Impeller Assembly (for use with probes) for Magnetically-Driven Proculture® Spinner Flask
fits flask, 6 L
Synonyme(s) :
equipment replacement accessory
About This Item
Code UNSPSC :
41121804
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
stainless steel shaft
Stérilité
non-sterile
Caractéristiques
fits flask 6 L
Fabricant/nom de marque
Corning 401577
Description générale
Stainless steel impeller shaft with dual bearing, modified impeller assembly for use in magnetically driven, 6L Proculture Spinner Flasks. Narrower impeller paddle design allows use with probes to create a small bioreactor.
Informations légales
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries