Z515612
Aldrich® evaporating flask, pear-shaped
capacity 500 mL, joint: ST/NS 24/40, non-coated, heavy wall
Synonyme(s) :
bottom flask, flask round, round flask
About This Item
Produits recommandés
Matériaux
borosilicate glass
long
pear-shaped
single-neck
Joint
joint: ST/NS24/40
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Fabricant/nom de marque
Büchi equivalent, 08758
Capacité
500 mL