Documents
BMSMW960012CM
Benchmark 12Channel Manifold
12-channel, manifold
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.01
Caractéristiques
12-channel manifold
Fabricant/nom de marque
Benchmark Scientific MW9600-12CM
Description générale
12 channel manifold for Accuris SmartWasher 96
Application
Washing microplates for ELISA assays