Merck
HomeSmall Molecule HPLCFingolimod HPLC Assay & Impurity Profiling (USP)
Fingolimod HPLC Assay & Impurity Profiling (USP)
Introduction
A rapid, accurate, and simple method was implemented for the total chromatographic purity analysis of Fingolimod Hydrochloride by High Performance Liquid Chromatography equipped with a Diode Array Detector. The experimental conditions follow guidelines, with minor allowed modifications, from the USP43-NF38 monograph methods for Fingolimod Hydrochloride Assay and Organic Impurities Profiling. Fingolimod hexyl homolog, Fingolimod heptyl homolog, Fingolimod Hydrochloride, Fingolimod nonyl homolog, Fingolimod decyl homolog, 3-Phenethyl Fingolimod analog, and 2-Phenethyl Fingolimod analog can be resolved with baseline separation utilizing the USP‘s 33-minute gradient method and a Purospher STAR RP-18 HPLC column (150 x 3.0 mm, 3 μm). A 0.1% solution of Phosphoric acid in water and acetonitrile were employed as the mobile phases for the gradient elution. Under applied conditions, system suitability criteria are met, and the method demonstrates good resolution/selectivity, reproducibility, and sensitivity.
Chemical Structures
Chemical structure diagram featuring a benzene ring connected to an eight-carbon chain with alternating single and double bonds, ending in an amine group (NH2) bonded to a carbon atom that also has two hydroxyl groups (OH), accompanied by hydrochloric acid (HCl).
Figure 1.Fingolimod HCl
Structural formula of a molecule featuring a benzene ring connected to a chain of carbon atoms. The chain includes various functional groups: an amine (NH2), two hydroxyl groups (OH), and a methyl group (CH3) at the opposite end of the benzene ring.
Figure 2.Fingolimod Hexyl Homolog
FingolimodHeptyl Homolog
Figure 3.FingolimodHeptyl Homolog
O-Acetyl Fingolimod
Figure 4.O-Acetyl Fingolimod
Fingolimod Nonyl Homolog
Figure 5.Fingolimod Nonyl Homolog
Fingolimod Decyl Homolog
Figure 6.Fingolimod Decyl Homolog
Fingolimod 3-Phenethyl Analog
Figure 7.Fingolimod 3-Phenethyl Analog
Fingolimod 2-Phenethyl Analog
Figure 8.Fingolimod 2-Phenethyl Analog
Experimental Conditions
ColumnPurospher® STAR RP-18 endcapped (3µm) Hibar RT 150x3.0 mmInjection Volume5 µL
DetectionUV @ 215 nm (DAD)Flow Rate0.8 mL/min
Mobile PhaseA: 0.1% Phosphoric Acid in H2O  B: AcetonitrileTemperatureColumn: 40 °C
Autosampler: 10 °C
GradientSee tablePressure135-300 bar
DiluentMobile Phase A:B (50:50)  
Test SolutionDissolve 15 mg of Fingolimod Hydrochloride CRS in 25 mL diluent (0.6 mg/mL).
System Suitability SolutionDissolve 15.0 mg of USP Fingolimod for System Suitability using 25 mL diluent (0.6 mg/mL).
Standard SolutionDilute 1.0 mL of the test solution to 100.0 mL using mobile phase further dilute 1.0 mL of this solution to 10.0 mL using the mobile phase (0.003 mg/mL).
Time (min)A (%)B (%)
08020
20595
23595
23.18020
338020
Chromatographic Data (System Suitability Solution)
PeaksCompoundRetention Time (min)RRTResolutionTailing factor
1Fingolimod Hexyl Homolog9.00.84-1.0
2Fingolimod Heptyl Homolog10.10.9411.61.1
3Fingolimod10.71.002.14.4
4O-Acetyl Fingolimod11.81.103.00.7
5Fingolimod Nonyl Homolog12.31.154.21.2
6Fingolimod Decyl Homolog13.51.269.81.4
73-Phenethyl Fingolimod Analog19.81.8542.10.8
82-Phenethyl Fingolimod Analog20.21.892.81.2
Specificity (System Suitability Solution)
PeaksCompoundRetention Time (min)RRTRRT Requirement (USP43-NF38)ResolutionResolution (USP43-NF38)TailingTailing (USP43-NF38)
1Fingolimod Hexyl Homolog9.00.840.82--1.0-
2Fingolimod Heptyl Homolog10.10.940.9311.6-1.1-
3Fingolimod10.71.001.002.1-4.4Not more than (NMT) 5
4O-Acetyl Fingolimod11.81.10-3.0-0.7-
5Fingolimod Nonyl Homolog12.31.151.134.21.2 (between peak 4 and 5)1.2-
6Fingolimod Decyl Homolog13.51.261.239.8-1.4-
73-Phenethyl Fingolimod Analog19.81.851.9742.1-0.8-
82-Phenethyl Fingolimod Analog20.21.892.002.80.8 (between peak 7 and 8)1.2-
Repeatability (System Suitability Solution)
PeaksCompoundArea Response (N=3)Standard DeviationRSD (%)RSD (%) (USP43-NF38)  
3Fingolimod5496.12.30.04NMT 0.73%