Merck
UHPLC Analysis of p-Coumaric Acid on Titan C18
UHPLC Analysis of p-Coumaric Acid on Titan C18 application for UHPLC
Materials
standard
Product No.
Description
Pricing
p-Coumaric acid
≥98.0% (HPLC)
CONDITIONS
column
Titan C18, 10 cm x 2.1 mm I.D., 1.9 μm particles (577124-U)
mobile phase
[A] 10 mM ammonium acetate, pH 7.0; [B] acetonitrile (70:30, A;B)
flow rate
0.3 mL/min
column temp.
35 °C
detector
MS, XIC m/z 163.15
injection
2 μL
sample
300 ng/mL each in acetonitrile:water (3:1)
Description
Legal Information
null