UHPLC Analysis of Methapyrilene and Tripelennamine on Titan C18
CONDITIONS
column
Titan C18, 10 cm x 2. 1 mm I.D., 1.9 um particles (577124-U)
mobile phase
[A] 10 mM ammonium acetate, pH 7.0; [B] acetonitrile (70:30, A;B)
flow rate
0.3 mL/min
column temp.
35 °C
detector
MS, XIC m/z 262.14 and 256.18
injection
2 μL
sample
300 ng/mL each in acetonitrile:water (3:1)
Description
Legal Information
null