Merck
LC/MS/MS Analysis of Vitamin D Metabolites on Ascentis® Express F5 (Monitored Ions)
LC/MS/MS Analysis of Vitamin D Metabolites on Ascentis® Express F5 (Monitored Ions) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express F5, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 2.1 mm
mobile phase component
Product No.
Description
Pricing
Ammonium formate
eluent additive for LC-MS, LiChropur, ≥99.0%
standard
Product No.
Description
Pricing
1α-Hydroxyvitamin D3
≥97.0% (HPLC)
3-epi-25-Hydroxyvitamin D3
98% (CP)
3-epi-25-Hydroxyvitamin D2 solution
100 μg/mL in ethanol, 98% (CP)
25-Hydroxyvitamin D2 solution
50 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
25-Hydroxyvitamin D3 solution
100 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
1α,25-Dihydroxyvitamin D3 solution
5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
1α,25-Dihydroxyvitamin D2 solution
5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
CONDITIONS
column
Ascentis Express  F5, 15 cm x 2.1 mm I.D., 2.7 μm particles (53571-U)
mobile phase
[A] 5 mM ammonium formate water; [B] methanol; (25:75, A:B)
flow rate
0.4 mL/min
pressure
5178 psi (357 bar)
column temp.
40 °C
detector
ESI(+), 100-1000 m/z
injection
2 μL, each compound 300 ng/mL in 40:60 (water:methanol)
Description
Analysis Note
The metabolic role and health implications of the various vitamin D isoforms are of current clinical interest. Therefore, it is important to have an analytical method that will resolve all of the known isoforms with necessary sensitivity and specificity.This application shows that a combination of Ascentis Express F5 with MS detection provided rapid resolution of seven such compounds. The highest grade LC-MS solvents were used to supply low background interference and low particulate contaminants for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany