HPLC Analysis of Proteins and Peptides on Proteomix® WCX, Comparison of Different Particle Sizes
![HPLC Analysis of Proteins and Peptides on Proteomix® WCX, Comparison of Different Particle Sizes application for HPLC](/deepweb/assets/sigmaaldrich/product/images/181/128/0f74bccc-1489-41c7-864b-4ed1a0f27584/800/0f74bccc-1489-41c7-864b-4ed1a0f27584.jpg)
Materials
analytical column
Product No.
Description
Pricing
Proteomix® WCX-NP10 Weak Cation Exchange Column
L × I.D. 5 cm × 4.6 mm, 10 μm particle size, stainless steelProteomix® WCX-NP1.7 Weak Cation Exchange Column
L × I.D. 5 cm × 4.6 mm, 1.7 μm particle size, stainless steelProteomix® WCX-NP3 Weak Cation Exchange Column
L × I.D. 5 cm × 4.6 mm, 3 μm particle size, stainless steelProteomix® WCX-NP5 Weak Cation Exchange Column
L × I.D. 5 cm × 4.6 mm, 5 μm particle size, stainless steelCONDITIONS
column
Proteomix WCX NP3, 5 cm x 4.6 mm I.D., 3 μm particles (Z777185)
column
Proteomix WCX-NP1.7, 5 cm x 4.6 mm I.D., 1.7 μm particles (Z777180)
column
Proteomix WCX-NP10, 5 cm x 4.6 mm, 10 μm particles (Z777169)
column
Proteomix WCX-NP5, 5 cm x 4.6 mm x 5 μm particles (Z777193)
mobile phase
(A) 20 mM sodium phosphate, pH 6.5, (B) A + 1.0 M NaCl
gradient
0 to 100% B in 25 min, 15 min prewash
flow rate
0.5 mL/min
column temp.
ambient
detector
UV, 214 nm
injection
5 μL
sample
Ribonuclease A, Cytochrome C and Lysozyme (1 mg/mL) min
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies