HPLC Analysis of Ovalbumin and BSA Charge Variants on Proteomix® SAX-NP3
![HPLC Analysis of Ovalbumin and BSA Charge Variants on Proteomix® SAX-NP3 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/382/421/45e2103e-38c5-4daf-8ac4-b1d176a23bb9/800/45e2103e-38c5-4daf-8ac4-b1d176a23bb9.jpg)
Materials
analytical column
Product No.
Description
Pricing
Zenix® SEC-300 Gel Filtration Column, 3 μm
L × I.D. 15 cm × 7.8 mm, 3 μm particle size, stainless steelCONDITIONS
column
Proteomix SAX-NP3, 5 cm x 4.6 mm I.D., 3 μm particles (Z777219)
mobile phase
20 mM Tris buffer, pH 8.0
gradient
0 - 0.3 M NaCl in 15 min
flow rate
0.5 mL/min
pressure
1600 psi (110 bar)
detector
UV, 214 nm
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies