GC Analysis of Ionic Liquid Test Mix on SLB®-IL100 (15 m x 0.10 mm I.D., 0.08 μm)
![GC Analysis of Ionic Liquid Test Mix on SLB®-IL100 (15 m x 0.10 mm I.D., 0.08 μm) suitable for GC](/deepweb/assets/sigmaaldrich/product/images/402/113/96d49b9b-47de-41c7-9e27-c2aaabd58c9b/800/96d49b9b-47de-41c7-9e27-c2aaabd58c9b.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL100, 15 m x 0.10 mm I.D., 0.08 μm (28882-U)
oven
90 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
hydrogen, 50 cm/sec
detector
0.5 μL 300:1 split
sample
Ionic Liquid Test Mix #1, each analyte at various concentrations in isooctane (N09000)
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany