GC Analysis of a 37-Component FAME Mix on SLB®-IL100
![GC Analysis of a 37-Component FAME Mix on SLB®-IL100 suitable for GC](/deepweb/assets/sigmaaldrich/product/images/205/357/f8c2347f-9c4e-4bb7-b967-922ae33793cc/800/f8c2347f-9c4e-4bb7-b967-922ae33793cc.jpg)
Materials
analytical column
Product No.
Description
Pricing
standard
Product No.
Description
Pricing
Supelco 37 Component FAME Mix
certified reference material, TraceCERT®, in dichloromethane (varied conc.), ampule of 1 mLCONDITIONS
column
SLB-IL100, 30 m x 0.25 mm I.D., 0.20 μm (28884-U)
oven
50 °C, 3.0 °C/min to 240 °C
inj. temp.
240 °C
detector
FID, 240 °C
carrier gas
helium, 40 cm/sec constant
injection
1 μL 50:1 split
sample
Supelco 37-Component FAME Mix (47885-U), analytes at concentrations indicated in methylene chloride
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany