Merck
GC Analysis of 1-Cyclohexylethylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX B-DP
GC Analysis of 1-Cyclohexylethylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX™ B-DP suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX B-DP Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
(R)-(−)-1-Cyclohexylethylamine
98%
(S)-(+)-1-Cyclohexylethylamine
98%
CONDITIONS
column
Astec CHIRALDEX B-DP, 30 m x 0.25 mm I.D., 0.12 μm (78023AST)
oven
130 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
sample
peak 1: S(+)-cyclohexylethylamine (N-TFA derivative)
peak 2: R(-)-cyclohexylethylamine (N-TFA derivative)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC