GC Analysis of FAMEs in Plasma on SUPELCOWAX 10 (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
CONDITIONS
column
SUPELCOWAX 10, 10 m × 0.10 mm I.D., 0.10 μm (25026-U)
oven
220 °C, 60 °C/min to 280 °C (1 min)
inj. temp.
280 °C
detector
FID, 280 °C
carrier gas
hydrogen, 120 cm/sec constant
injection
0.5 μL, 30:1 split
sample
plasma FAMEs in hexane
Description
Legal Information
null