Merck
  • Determination of volatile organic compounds in contaminated air using semipermeable membrane devices.
Determination of volatile organic compounds in contaminated air using semipermeable membrane devices.
Talanta (2010-02-16)
Saray Ly-Verdú, Francesc A Esteve-Turrillas, Agustín Pastor, Miguel de la Guardia
ABSTRACT
Semipermeable membrane devices (SPMDs) were evaluated as passive samplers for the determination of 26 volatile organic compounds (VOCs) in contaminated air of occupational environments. A direct methodology based on the use of head-space-gas chromatography-mass spectrometry (HS-GC-MS) was developed for VOCs determinations in SPMDs, without any sample pre-treatment and avoiding the use of solvents. A desorption temperature of 150 degrees C for 10 min was sufficient for a sensitive VOCs determination providing limits of detection in the range of 15 ng SPMD(-1) for 21 of 26 studied compounds. Linear and equilibrium uptake models were established for each VOC from compound isotherms. Highly volatile compounds were slightly absorbed and moderately volatile compounds were strongly absorbed by SPMDs. This study is the first precedent of the use of SPMDs for the simultaneous sampling of a wide number of VOCs. The use of SPMDs is a simple and low cost alternative to ordinary sampling devices such as Radiello diffusive samplers or badge-type solid-phase supports.
Product Description
radiello BTEX/VOC Starter Kit, CS2 Desorption
radiello BTEX/VOC Starter Kit, Thermal Desorption
radiello Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
radiello Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
radiello Diffusive Bodies, permeative, configured for anaesthetic gases and vapors, pk of 20
radiello Filtration Kit, pk of 20
Supelco
radiello Aldehyde Calibration Standard, 50 μg/mL each component in acetonitrile, pkg of 10 mL
radiello Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
radiello 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
Supelco
radiello BTEX Calibration Kit (CS2 Desorption)
radiello On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
radiello Clips, suspending Radiello triangular support plate, pk of 20
radiello Diffusive Bodies, white, configured for general use, pk of 20
radiello On-Field Thermometer, Reader, pk of 1
Supelco
radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
radiello Empty Storage Tubes, glass (with stopper), volume 2.8 mL, pk of 100
radiello Hydrogen Sulfide Starter Kit
Supelco
radiello BTEX Calibration Kit (Thermal Desorption)
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (CS2 Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20
radiello Barcode Labels, pk of 198
radiello Cartridge Adsorbents, for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (thermal desorption), matrix SS net (3 x 8 μm, 4.8 mm diam.), Carbograph, pk of 20
radiello Cartridge Adsorbents, for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL®, pk of 20
radiello Anesthetic Gases and Vapor Sampler
radiello Cartridge Adsorbents, for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20
radiello Replacement Mounting Strips, for use with Radiello Outdoor Shelter, pk of 100
radiello Outdoor Shelter, pk of 10
radiello Vertical Adapter, threaded for standard use, pk of 20
radiello Triangular Support Plate, pk of 20