Merck
  • [Simultaneous determination of N-methylcarbamate pesticides and their metabolites in agricultural products].
[Simultaneous determination of N-methylcarbamate pesticides and their metabolites in agricultural products].
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2002-09-20)
Maki Kobayashi, Toshihiro Nagayama, Ichiro Takano, Yasuhiro Tamura, Naoko Kimura, Kyoko Kitayama, Kazuo Saito
ABSTRACT
We investigated the determination of N-methylcarbamate pesticides and their metabolites in agricultural products by HPLC with post-column fluorescence detection after clean-up with an SPE cartridge. The homogenate of agricultural products was extracted with acetone. The crude extract was partitioned between 5% sodium chloride solution and dichloromethane, and the dichloromethane layer was concentrated to dryness. The residue was dissolved with a mixture of acetone and n-hexane, and purified by using Supelclean ENVI-Carb SPE, Bond Elut Extraction Cartridge PSA and SAX in series with a mixture of acetone and n-hexane. N-Methylcarbamate pesticide was analyzed by HPLC with post-column reaction and fluoresce detection. N-Methylcarbamate pesticide in citrus fruits and various kinds of agricultural products could be analyzed accurately by the presented method. Recoveries of N-methylcarbamate pesticides added to several agricultural products at the level of 0.10 ppm were mostly in the range of 60-110%. The limit of detection was 0.005 ppm.
Product Description
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS