Merck
  • Determination of six polyether antibiotic residues in foods of animal origin by solid phase extraction combined with liquid chromatography-tandem mass spectrometry.
Determination of six polyether antibiotic residues in foods of animal origin by solid phase extraction combined with liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2016-03-19)
Jing Ha, Ge Song, Lian-Feng Ai, Jian-Chen Li
ABSTRACT
A new method using solid phase extraction (SPE) combined with liquid chromatography-tandem mass spectrometry (LC-MS/MS) has been developed for the determination of six polyether antibiotics, including lasalocid, salinomycin, monensin, narasin, madubamycin and nigericin residues, in foods of animal origin. The samples were extracted with acetonitrile and purified by ENVI-Carb SPE columns after comparing the impurity effect and maneuverability of several SPE cartridges. Subsequently, the analytes were separated on a Hypersil Gold column (2.1×150mm, 5μm) and analyzed by MS/MS detection. The limit of quantization (LOQ) for milk and chicken was 0.4μg/kg, and for chicken livers and eggs, it was 1μg/kg. The linearity was satisfactory with a correlation coefficient of >0.9995 at concentrations ranging from 2 to 100μg/L. The average recoveries of the analytes fortified at three levels ranged from 68.2 to 114.3%, and the relative standard deviations ranged from 4.5 to 12.1%. The method was suitable for quantitative analysis and confirmation of polyether antibiotic residues in foods of animal origin.
Product Description
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01