Merck
  • Significance of morphometric, DNA cytometric features, and other prognostic markers on survival of endometrial cancer patients in northern Norway.
Significance of morphometric, DNA cytometric features, and other prognostic markers on survival of endometrial cancer patients in northern Norway.
International journal of gynecological cancer : official journal of the International Gynecological Cancer Society (2002-03-27)
A Ørbo, M Rydningen, B Straume, S Lysne
ABSTRACT
The objective of this study was to evaluate the prognostic value of nuclear morphometric features and DNA ploidy by flow cytometry next to depth of myometrial invasion and vascular invasion in endometrial cancer of all FIGO stages. A total of 123 women (103 FIGO stage I, eight stage II, and 12 stage III and IV) from northern Norway were studied. The follow-up period was between 7 and 19 years. The median age of patients was 62 years. The primary surgery was performed in the University Hospital of Tromso or in the seven different reference hospitals in the northern part of Norway after an endometrial cancer diagnosis. The histologic, morphometric,flow cytometric and immunohistochemical investigations were based on archival paraffin-embedded material. The information regarding the follow-up data and clinical information were obtained from the medical records. Thirteen (10.6%) patients from the entire group (all stages) but only three (2.7%) of the FIGO stage I and II patients died from locally recurrent or metastatic disease. FIGO substage (P = 0.0006; odds ratio [OR] =16.44, 95% confidence interval [CI] = 3.36-80.45), vascular invasion (P =0.01, OR = 6.42, CI = 1.57-26.34) and nuclear size (P = 0.025, OR = 1.3,CI = 1.05-1.61) were independently correlated with recurrence in a multivariate analysis but histologic grade and DNA ploidy were not. Vascular invasion was poorly reproducible both between and within the same observer, however. In this retrospective study of all stages of endometrial carcinoma with long follow-up periods the primary tumor characteristics nuclear perimeter and FIGO stage were of prognostic significance in addition to the poorly reproducible vessel invasion.
Product Description
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate