Merck
  • Evaluation of chromosome aberration and micronucleus frequencies in blood lymphocytes of workers exposed to low concentrations of benzene.
Evaluation of chromosome aberration and micronucleus frequencies in blood lymphocytes of workers exposed to low concentrations of benzene.
Mutation research. Genetic toxicology and environmental mutagenesis (2014-10-26)
Piero Lovreglio, Francesca Maffei, Mariella Carrieri, Maria N D'Errico, Ignazio Drago, Patrizia Hrelia, Giovanni B Bartolucci, Leonardo Soleo
ABSTRACT
The frequency of chromosome aberrations (CA) and micronuclei (MN) was investigated in the peripheral lymphocytes of workers occupationally exposed to low or very low concentrations of benzene. The study included 43 exposed workers (all males), namely 19 fuel-tanker drivers and 24 filling-station attendants, and 31 male subjects with no occupational exposure to the toxicant (controls). Benzene exposure was verified by means of environmental monitoring with passive personal samplers (Radiello(®)), and through biological monitoring, i.e. by measurement of urinary trans,trans-muconic acid, S-phenylmercapturic acid and benzene. The frequency of CA and MN in peripheral lymphocytes was determined according to standard procedures. Exposure to benzene was found to be significantly higher for fuel-tanker drivers (median 246.6 μg/m(3)) than for filling-station attendants (median 19.9 μg/m(3)). Both groups had significantly higher exposure than controls (median 4.3 μg/m(3)). No increased frequency of CA and MN was observed in either fuel-tanker drivers or filling-station attendants compared with controls. In all subjects examined as a single group, the frequency of MN was significantly dependent on age. Only in the fuel-tanker drivers was the frequency of MN found to depend not only on age, but also on exposure to benzene. In conclusion, the frequency of MN, but not of CA, could be influenced by exposure to benzene concentrations of up to one order of magnitude lower than the threshold limit value (time-weighted average).
Product Description
Supelco
radiello BTEX Calibration Kit (Thermal Desorption)
radiello On-Field Thermometer, Reader, pk of 1
radiello Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
radiello Empty Storage Tubes, glass (with stopper), volume 2.8 mL, pk of 100
radiello Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
radiello Filtration Kit, pk of 20
radiello Clips, suspending Radiello triangular support plate, pk of 20
radiello On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
radiello BTEX/VOC Starter Kit, CS2 Desorption
radiello Diffusive Bodies, permeative, configured for anaesthetic gases and vapors, pk of 20
radiello Diffusive Bodies, white, configured for general use, pk of 20
radiello BTEX/VOC Starter Kit, Thermal Desorption
radiello 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
Supelco
radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
Supelco
radiello BTEX Calibration Kit (CS2 Desorption)
radiello Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
Supelco
radiello Aldehyde Calibration Standard, 50 μg/mL each component in acetonitrile, pkg of 10 mL
radiello Hydrogen Sulfide Starter Kit
radiello Vertical Adapter, threaded for standard use, pk of 20
radiello Cartridge Adsorbents, for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL®, pk of 20
radiello Cartridge Adsorbents, for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20
radiello Replacement Mounting Strips, for use with Radiello Outdoor Shelter, pk of 100
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (CS2 Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20
radiello Anesthetic Gases and Vapor Sampler
radiello Barcode Labels, pk of 198
radiello Outdoor Shelter, pk of 10
radiello Cartridge Adsorbents, for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (thermal desorption), matrix SS net (3 x 8 μm, 4.8 mm diam.), Carbograph, pk of 20
radiello Triangular Support Plate, pk of 20